Anti-CD72 Mouse Monoclonal Antibody [clone: 10B1957]

Lieferant: United States Biological
USBIC2428-41VEA 594 EUR
USBIC2428-41V
Anti-CD72 Mouse Monoclonal Antibody [clone: 10B1957]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100; Immunochemistry: 1:50

Type: Primary
Antigen: CD72 (CD72 molecule)
Clonality: Monoclonal
Clone: 10B1957
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR