Anti-TFRC Mouse Monoclonal Antibody [clone: 8J49]
Lieferant: United States Biological
USBIC2426-28DEA 299 EUR
USBIC2426-28D
Anti-TFRC Mouse Monoclonal Antibody [clone: 8J49]
Antikörper
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: Monoclonal
Clone: 8J49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human