Anti-TFRC Mouse Monoclonal Antibody [clone: 5F54]

Lieferant: United States Biological
USBIC2426-19EA 368 EUR
USBIC2426-19
Anti-TFRC Mouse Monoclonal Antibody [clone: 5F54]
Antikörper
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: Monoclonal
Clone: 5F54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR