Anti-CD69 Mouse Monoclonal Antibody [clone: BL-Ac/p26]

Lieferant: United States Biological
USBIC2424-05JEA 0 EUR
USBIC2424-05J
Anti-CD69 Mouse Monoclonal Antibody [clone: BL-Ac/p26]
Antikörper
Type: Primary
Antigen: CD69 (CD69 molecule)
Clonality: Monoclonal
Clone: BL-Ac/p26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR