Anti-CD68 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B1906]

Lieferant: United States Biological
USBIC2423-05P9EA 748 EUR
USBIC2423-05P9
Anti-CD68 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B1906]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: Monoclonal
Clone: 10B1906
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR