Anti-FCGR1A Mouse monoclonal antibody [clone: 7H102]

Lieferant: United States Biological
USBIC2419-06GEA 370 EUR
USBIC2419-06G
Anti-FCGR1A Mouse monoclonal antibody [clone: 7H102]
Antikörper
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: Monoclonal
Clone: 7H102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR