Anti-SELP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H186]

Lieferant: United States Biological
USBIC2415-55HEA 414 EUR
USBIC2415-55H
Anti-SELP Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H186]
Antikörper
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: SELP (selectin P (granule membrane protein 140kDa, antigen CD62))
Clonality: Monoclonal
Clone: 7H186
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR