Anti-SELL Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC2415-31M-BIOTEA 678 EUR
USBIC2415-31M-BIOT
Anti-SELL Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: SELL (selectin L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR