Anti-SELL Mouse Monoclonal Antibody [clone: 5F51]

Lieferant: United States Biological
USBIC2415-15EEA 309 EUR
USBIC2415-15E
Anti-SELL Mouse Monoclonal Antibody [clone: 5F51]
Antikörper
Type: Primary
Antigen: SELL (selectin L)
Clonality: Monoclonal
Clone: 5F51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR