Anti-SELL Mouse Monoclonal Antibody (Biotin) [clone: 7H203]

Lieferant: United States Biological
USBIC2415-15BEA 520 EUR
USBIC2415-15B
Anti-SELL Mouse Monoclonal Antibody (Biotin) [clone: 7H203]
Antikörper
Type: Primary
Antigen: SELL (selectin L)
Clonality: Monoclonal
Clone: 7H203
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR