Anti-SELE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6D40]

Lieferant: United States Biological
USBIC2415-06CEA 539 EUR
USBIC2415-06C
Anti-SELE Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6D40]
Antikörper
Type: Primary
Antigen: SELE (selectin E)
Clonality: Monoclonal
Clone: 6D40
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR