Anti-ITGB3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC2414-20KEA 457 EUR
USBIC2414-20K
Anti-ITGB3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR