Anti-CD60b Mouse Monoclonal Antibody (Biotin) [clone: 7H226]

Lieferant: United States Biological
USBIC2413-79AEA 0 EUR
USBIC2413-79A
Anti-CD60b Mouse Monoclonal Antibody (Biotin) [clone: 7H226]
Antikörper
Type: Primary
Antigen: CD60b
Clonality: Monoclonal
Clone: 7H226
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR