Anti-CD60 Mouse Monoclonal Antibody [clone: M-T6004]

Lieferant: United States Biological
USBIC2413-75AEA 900 EUR
USBIC2413-75A
Anti-CD60 Mouse Monoclonal Antibody [clone: M-T6004]
Antikörper
Type: Primary
Antigen: CD60
Clonality: Monoclonal
Clone: M-T6004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR