Anti-CD59 Mouse Monoclonal Antibody [clone: 10B1845]

Lieferant: United States Biological
USBIC2413-21TEA 521 EUR
USBIC2413-21T
Anti-CD59 Mouse Monoclonal Antibody [clone: 10B1845]
Antikörper
Recommended dilution: Flow Cytometry: 10 ?g/ml;

Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: 10B1845
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR