Anti-CD59 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H165]

Lieferant: United States Biological
USBIC2413-21REA 0 EUR
USBIC2413-21R
Anti-CD59 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H165]
Antikörper
Recommended dilution: Flow Cytometry: 1:50
Do not freeze!

Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: 7H165
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR