Anti-CD58 Mouse Monoclonal Antibody [clone: 7H219]

Lieferant: United States Biological
USBIC2412-06JEA 370 EUR
USBIC2412-06J
Anti-CD58 Mouse Monoclonal Antibody [clone: 7H219]
Antikörper
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: Monoclonal
Clone: 7H219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR