Anti-NCAM1 Mouse Monoclonal Antibody [clone: 7H153]

Lieferant: United States Biological
USBIC2410-55CEA 370 EUR
USBIC2410-55C
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 7H153]
Antikörper
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 7H153
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR