Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K210]

Lieferant: United States Biological
USBIC2410-50BEA 405 EUR
USBIC2410-50B
Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K210]
Antikörper
Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: Monoclonal
Clone: 5K210
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR