Anti-ICAM1 Mouse Monoclonal Antibody [clone: 6A507]

Lieferant: United States Biological
USBIC2409-47EA 569 EUR
USBIC2409-47
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 6A507]
Antikörper
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: Monoclonal
Clone: 6A507
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR