Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]

Lieferant: United States Biological
USBIC2409-11MEA 473 EUR
USBIC2409-11M
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]
Antikörper
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: Monoclonal
Clone: 15.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR