Anti-ICAM1 Mouse Monoclonal Antibody (Biotin) [clone: 1A29]

Lieferant: United States Biological
USBIC2409-11C3EA 0 EUR
USBIC2409-11C3
Anti-ICAM1 Mouse Monoclonal Antibody (Biotin) [clone: 1A29]
Antikörper
Recommended dilution: Flow Cytometry:1:5;

Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: Monoclonal
Clone: 1A29
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR