Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1532]

Lieferant: United States Biological
USBIC2407-50J2EA 541 EUR
USBIC2407-50J2
Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1532]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD52 (CD52 molecule)
Clonality: Monoclonal
Clone: 3H1532
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR