Anti-ITGA5 Mouse Monoclonal Antibody [clone: 5K288]

Lieferant: United States Biological
USBIC2405-01HEA 443 EUR
USBIC2405-01H
Anti-ITGA5 Mouse Monoclonal Antibody [clone: 5K288]
Antikörper
Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: Monoclonal
Clone: 5K288
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR