Anti-ITGA4 Rat Monoclonal Antibody [clone: 5K278]

Lieferant: United States Biological
USBIC2404-65AXEA 394 EUR
USBIC2404-65AX
Anti-ITGA4 Rat Monoclonal Antibody [clone: 5K278]
Antikörper
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: Monoclonal
Clone: 5K278
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR