Anti-ITGA4 Mouse Monoclonal Antibody [clone: 10B1722]

Lieferant: United States Biological
USBIC2404-46KEA 594 EUR
USBIC2404-46K
Anti-ITGA4 Mouse Monoclonal Antibody [clone: 10B1722]
Antikörper
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: Monoclonal
Clone: 10B1722
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR