Anti-CD48 Mouse Monoclonal Antibody [clone: 7H129/ 0.N.140]
Lieferant: United States Biological
USBIC2403-15CEA 473 EUR
USBIC2403-15C
Anti-CD48 Mouse Monoclonal Antibody [clone: 7H129/ 0.N.140]
Antikörper
Type: Primary
Antigen: CD48 (CD48 molecule)
Clonality: Monoclonal
Clone: 7H129/ 0.N.140
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human