Anti-CD48 Hamster Monoclonal Antibody

Lieferant: United States Biological
USBIC2403-09W-50EA 246 EUR
USBIC2403-09W-50
Anti-CD48 Hamster Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CD48 (CD48 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR