Anti-CD48 Mouse Monoclonal Antibody [clone: 3H1465]

Lieferant: United States Biological
USBIC2403-09REA 365 EUR
USBIC2403-09R
Anti-CD48 Mouse Monoclonal Antibody [clone: 3H1465]
Antikörper
Recommended dilution: Flow Cytometry: 1:100-1:200;

Type: Primary
Antigen: CD48 (CD48 molecule)
Clonality: Monoclonal
Clone: 3H1465
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR