Anti-CD46 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B1688]

Lieferant: United States Biological
USBIC2401-02F1EA 666 EUR
USBIC2401-02F1
Anti-CD46 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B1688]
Antikörper
Type: Primary
Antigen: CD46 (CD46 molecule, complement regulatory protein)
Clonality: Monoclonal
Clone: 10B1688
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR