Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: 104-2]
Lieferant: United States Biological
USBIC2400-91GEA 540 EUR
USBIC2400-91G
Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5.5®) [clone: 104-2]
Antikörper
Type: Primary
Antigen: CD45.2 (CD45.2)
Clonality: Monoclonal
Clone: 104-2
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse