Anti-CD45.1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A291]

Lieferant: United States Biological
USBIC2400-91EA 374 EUR
USBIC2400-91
Anti-CD45.1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A291]
Antikörper
Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: Monoclonal
Clone: 6A291
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR