Anti-CD45.1 Mouse Monoclonal Antibody (Cy5®)) [clone: A20]

Lieferant: United States Biological
USBIC2400-83FEA 416 EUR
USBIC2400-83F
Anti-CD45.1 Mouse Monoclonal Antibody (Cy5®)) [clone: A20]
Antikörper
Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: Monoclonal
Clone: A20
Conjugation: Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR