Anti-CD45RC Rat Monoclonal Antibody [clone: 5K258]
Lieferant: United States Biological
USBIC2400-61AEA 499 EUR
USBIC2400-61A
Anti-CD45RC Rat Monoclonal Antibody [clone: 5K258]
Antikörper
Type: Primary
Antigen: CD45RC
Clonality: Monoclonal
Clone: 5K258
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM Kappa
Reactivity: Mouse