Anti-CD45RC Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K33]

Lieferant: United States Biological
USBIC2400-59MEA 273 EUR
USBIC2400-59M
Anti-CD45RC Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K33]
Antikörper
Type: Primary
Antigen: CD45RC
Clonality: Monoclonal
Clone: 5K33
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR