Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F42/5F5]

Lieferant: United States Biological
USBIC2400-44JEA 540 EUR
USBIC2400-44J
Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F42/5F5]
Antikörper
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: Monoclonal
Clone: 5F42/5F5
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a+IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR