Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H108]

Lieferant: United States Biological
USBIC2400-35D2EA 414 EUR
USBIC2400-35D2
Anti-CD45RA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H108]
Antikörper
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: Monoclonal
Clone: 7H108
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR