Anti-CD45RA Mouse Monoclonal Antibody [clone: HI100]

Lieferant: United States Biological
USBIC2400-30LEA 304 EUR
USBIC2400-30L
Anti-CD45RA Mouse Monoclonal Antibody [clone: HI100]
Antikörper
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: Monoclonal
Clone: HI100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR