Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: 5K224]

Lieferant: United States Biological
USBIC2400-05CEA 377 EUR
USBIC2400-05C
Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: 5K224]
Antikörper
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: 5K224
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR