Anti-PTPRC Mouse Monoclonal Antibody [clone: 5K237]

Lieferant: United States Biological
USBIC2399-14CEA 261 EUR
USBIC2399-14C
Anti-PTPRC Mouse Monoclonal Antibody [clone: 5K237]
Antikörper
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: 5K237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR