Anti-PTPRC Mouse Monoclonal Antibody [clone: 3H1378]

Lieferant: United States Biological
USBIC2399-07GEA 525 EUR
USBIC2399-07G
Anti-PTPRC Mouse Monoclonal Antibody [clone: 3H1378]
Antikörper
Recommended dilution: Flow Cytometry: 1:10-1:50; Immunochemistry: 1:100-1:1000;

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: 3H1378
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR