Anti-CD44 Rat Monoclonal Antibody [clone: 5K255]

Lieferant: United States Biological
USBIC2398-69DEA 362 EUR
USBIC2398-69D
Anti-CD44 Rat Monoclonal Antibody [clone: 5K255]
Antikörper
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 5K255
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR