Anti-CD44V3-10 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC2398-39WEA 580 EUR
USBIC2398-39W
Anti-CD44V3-10 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 50 ?g/ml

Type: Primary
Antigen: CD44V3-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR