Anti-CD44V3 Mouse Monoclonal Antibody (Biotin) [clone: 6D31]

Lieferant: United States Biological
USBIC2398-39EEA 409 EUR
USBIC2398-39E
Anti-CD44V3 Mouse Monoclonal Antibody (Biotin) [clone: 6D31]
Antikörper
Type: Primary
Antigen: CD44V3
Clonality: Monoclonal
Clone: 6D31
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR