Anti-CD44 Mouse Monoclonal Antibody [clone: 5K99]
Lieferant: United States Biological
USBIC2398-14EA 473 EUR
USBIC2398-14
Anti-CD44 Mouse Monoclonal Antibody [clone: 5K99]
Antikörper
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 5K99
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken