Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-85]

Lieferant: United States Biological
USBIC2398-06K1EA 0 EUR
USBIC2398-06K1
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-85]
Antikörper
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: MEM-85
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR