Anti-CD44 Mouse Monoclonal Antibody [clone: 6D26]

Lieferant: United States Biological
USBIC2398-05JEA 499 EUR
USBIC2398-05J
Anti-CD44 Mouse Monoclonal Antibody [clone: 6D26]
Antikörper
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 6D26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR