Anti-CD44 Mouse Monoclonal Antibody [clone: SPM521]

Lieferant: United States Biological
USBIC2398-02UEA 0 EUR
USBIC2398-02U
Anti-CD44 Mouse Monoclonal Antibody [clone: SPM521]
Antikörper
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: SPM521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR