Anti-CD44 Rat Monoclonal Antibody [clone: 10B1583]

Lieferant: United States Biological
USBIC2398-01TEA 0 EUR
USBIC2398-01T
Anti-CD44 Rat Monoclonal Antibody [clone: 10B1583]
Antikörper
Recommended dilution: Flow Cytometry: 1:20-1:50

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 10B1583
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Bovine, Cat, Dog, Horse, Human, Monkey, Mouse, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR