Anti-CD44 Mouse Monoclonal Antibody [clone: 10B1581]

Lieferant: United States Biological
USBIC2398-01NEA 668 EUR
USBIC2398-01N
Anti-CD44 Mouse Monoclonal Antibody [clone: 10B1581]
Antikörper
Recommended dilution: Flow Cytometry: 1:25-1:100; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 10B1581
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR